Ammonium chloride ≥98%, TECHNICAL
Catalog # 21224.368
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bucket
- Pk:5 kg
- Seq:4114
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
About this item
Synonyms: Ammonium muriate, Salmiac
:
:53.49 g/mol
Merck Index:13,00510
Melting Pt:335 °C
Density:1.5256 g/cm³ (20 °C)
EINECS:235-186-4
REACH:01-2119487950-27
:GHS07
Storage Temperature:Ambient
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420