Order Entry
Austria
ContactUsLinkComponent
Ammonium chloride, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Ammonium chloride, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Ammonium chloride, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    372 g/l (20 °C)
  • Melting Point:
    335 °C (sublimed)
  • Molar Mass:
    53.49 g/mol
  • Bulk Density:
    500 kg/m3
  • Vapor Pressure:
    1.3 hPa (30 °C)
  • Density:
    1.52 g/cm3 (20 °C)
  • pH:
    4.5 - 5.5 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    400 °C

Specification Test Results

Related Information

Frequently Bought Together