Order Entry
Austria
ContactUsLinkComponent
Tetrabutylammonium hydrogen sulphate ≥98% for synthesis
Irritant
Tetrabutylammonium hydrogen sulphate ≥98% for synthesis
Catalog # 28529.186
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Tetrabutylammonium hydrogen sulphate ≥98% for synthesis
Catalog # 28529.186
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Melting point:
    168 - 174 °C

Specification Test Results

About this item

 :
 :339.54 g/mol
Melting Pt:169…172 °C
Density:1.01 g/cm³ (25 °C)
EINECS:251-068-5
 :GHS07
CAS Number:32503-27-8
Formula:(C₄H₉)₄NHSO₄
MDL Number:MFCD00011637