Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes for Tubes

Storage Boxes for Tubes

Supplier:   Simport Scientific
Storage Boxes for Tubes
Supplier:  Simport Scientific
Certificates
Storage Boxes for Tubes
Storage Boxes for Tubes

4 Options Available Below

Product Details & Documents

Cover is made of PS and base made of high impact PS.

  • Stackable
  • Colour coded grid
  • Air vents minimising condensation

These 5 ml tube storage boxes are designed to store samples between –90 to +80 °C.

Printed with a series of numbered squares and the surface accepts the writing with markers by facilitating inventory control.

Transparent cover that allows the user to see the contents of the box and is keyed to the base to prevent misalignment.

Product Family Options

Product Information

  • No. of Holes
    Colour
    For tubes
    Availability
    Price
  • 25 (5×5)
    Blue
    5 ml
    Log in for availability
    Log in for availability
    1 * 18 ST
    1 * 18 ST - 313,00 €
    313,00 €
    1 * 18 ST
       313,00 €

  • 25 (5×5)
    Green
    5 ml
    Log in for availability
    Log in for availability
    1 * 18 ST
    1 * 18 ST - 313,00 €
    313,00 €
    1 * 18 ST
       313,00 €

  • 25 (5×5)
    Pink
    5 ml
    Log in for availability
    Log in for availability
    1 * 18 ST
    1 * 18 ST - 313,00 €
    313,00 €
    1 * 18 ST
       313,00 €

  • 25 (5×5)
    Yellow
    5 ml
    Log in for availability
    Log in for availability
    1 * 18 ST
    1 * 18 ST - 313,00 €
    313,00 €
    1 * 18 ST
       313,00 €