Tetrabutylammonium fluoride trihydrate for synthesis, Sigma-Aldrich®
: Merck
:87749-50-6
:
:C₁₆H₃₆FN·3H₂O
:MFCD00149981
:315.51 g/mol
:62…63 °C
:Ambient
:207-057-2
:2811
:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
- Solubility:(20 °C) soluble
- Melting Point:62 - 63 °C
- Molar Mass:315.49 g/mol
- Synonyms:TBAF
- Hill Formula:C16H36FN * 3H2O
- Chemical Formula:(CH3CH2CH2CH2)4NF * 3H2O
Frequently Bought Together