Tetramethylammonium chloride, Sigma-Aldrich®
Catalog # SIAL87718-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:109.60 g/mol
Melting Pt:> 300 °C
Density:1.169 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-880-8
UN:2811
ADR:6.1,III
:GHS06
CAS Number:75-57-0
Formula:(CH₃)₄NCl
MDL Number:MFCD00011628