Tetramethylammonium chloride, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:75-57-0
:
Formula:(CH₃)₄NCl
MDL Number:MFCD00011628
:109.60 g/mol
Melting Pt:> 300 °C
Density:1.169 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:200-880-8
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:2811
:GHS06
Storage Temperature:Ambient
Frequently Bought Together