Tetramethylammonium chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-57-0
:
Formula:(CH₃)₄NCl
MDL Number:MFCD00011628
:109.60 g/mol
Melting Pt:>300 °C
Density:1.17
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-880-8
UN:2811
ADR:6.1,II
:GHS06
:GHS07
Related Information
Frequently Bought Together