Tetrabutylammonium bromide ≥99.0% (by argentometric titration) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1643-19-2
:
Formula:C₁₆H₃₆BrN
MDL Number:MFCD00011633
Density:1.148 g/cm³ (20 °C)
Molecular Weight:322.37 g/mol
Melting Pt:100…103 °C
:M-Clarity™ quality level = MQ200
EINECS:216-699-2
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (argentometric):≥ 99.0 %
- Identity (IR):passes test
- Molar Mass:322.37 g/mol
- Bulk Density:700 kg/m3
- Density:1.15 g/cm3
Specification Test Results
Frequently Bought Together