ContactUsLinkComponent
Burette roller clamp, epoxy-coated steel
Burette roller clamp, epoxy-coated steel
Catalog # JUCHVL04512
undefined
Burette roller clamp, epoxy-coated steel
Catalog # JUCHVL04512
Specifications
  • For:
    One burette
  • Spanning width:
    0 – 30 mm
  • Material:
    Steel, epoxy coating
  • Pk:
    1
Specifications
About this item
Made of epoxy coated with one boss.