Order Entry
Austria
ContactUsLinkComponent
 
Buffer solution, pH 12,454
Catalog # APPC277089.1209
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Buffer solution, pH 12,454
Catalog # APPC277089.1209
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Reference temperature:
    25 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    12,454

Specifications

About this item

These buffer solutions are useful for the calibration of high precision laboratory pH meters.

  • pH (30 °C): 12,289 ±0,01
  • pH (25 °C): 12,454 ±0,01
  • pH (20 °C): 12,627 ±0,01
  • pH (10 °C): 12,810 ±0,01
Certifications: Standardised, according to DIN19266.