Tributyltin hydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Tri-n-butyltin hydride
CAS Number:688-73-3
:
Formula:C₁₂H₂₈Sn
MDL Number:MFCD00009416
:291.06 g/mol
Boiling Pt:112…114 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:109 °C
UN:2788
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:211-704-4
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Hill Formula:C₁₂H₂₈Sn
- Molar Mass:291.06 g/mol
- Boiling point:112 - 114 °C (11 hPa)
- Density:1.10 g/cm³ (20 °C)
- Flash point:109 °C
- Ignition temperature:250 °C
- Vapor pressure:5 hPa (20 °C)
- Refractive Index:1.4726 (20 °C, 589 nm)
- Storage class:6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
- WGK:WGK 3 highly water endangering
- undefined:R Phrase
- R 21-25-36/38-48/23/25-50/53:S Phrase
Specification Test Results
Frequently Bought Together