
Trioctyltin chloride
Catalog # EHERC17892000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:493.83 g/mol
EINECS:219-969-8
:GHS07
CAS Number:2587-76-0
Formula:C₂₄H₅₁ClSn