Order Entry
Austria
ContactUsLinkComponent
 
Dimethyldichlorosilane 99+%
 
undefined
Dimethyldichlorosilane 99+%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4030 to 1.4060 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Absorption Ratio P-15/P+15:
    1.21 to 1.95
  • BSC Certification:
    Passes

 

Frequently Bought Together