
Methyltriethoxysilane for synthesis, Sigma-Aldrich®
8.18819.0250
: Merck
:
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
- Solubility:(20 °C) (decomposition)
- Molar Mass:178.30 g/mol
- Boiling Point:142 °C (1013 hPa)
- Vapor Pressure:13.3 hPa (20 °C)
- Flash Point:33 °C
- Refractive Index:1.3868 (20 °C, 589 nm)
- Density:0.89 g/cm3 (20 °C)
- Ignition Point:220 °C
:
:178.3 g/mol
:143…145 °C (1013 hPa)
:–50 °C
:0.89 g/cm³ (20 °C)
:30 °C
:Refrigerator
:217-983-9
:1993
:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:2031-67-6
:C₇H₁₈O₃Si
:MFCD00009066