Order Entry
Austria
ContactUsLinkComponent
Carbon disulphide ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Flammable
Health
Irritant
Carbon disulphide ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates
undefined
Flammable
Health
Irritant
Carbon disulphide ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.03 %
  • Absorbance at 750 nm:
    Max. 0.01
  • Absorbance 500 nm:
    Max. 0.01
  • Absorbance at 410 nm:
    Max. 0.05
  • Absorbance at 403 nm:
    Max. 0.10
  • Absorbance at 394 nm:
    Max. 0.25
  • Absorbance at 388 nm:
    Max. 0.50
  • Absorbance at 386 nm:
    Max. 1.0
  • Autoignition Temperature:
    212 °F
  • Explosion Limit:
    50 %
  • Vapor Density:
    2.67 (vs air)

Specification Test Results

Related Information

Frequently Bought Together