Silicon carbide 400 grinding compound
Catalog # 39800.LA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:8425
- Environmentally Preferable:
- Pk Info:2oz (57g)
Specifications
About this item
CAS Number:409-21-2
:
Formula:SiC
MDL Number:MFCD00049531
:40.1 g/mol
Melting Pt:2700 °C
Density:3.16
EINECS:206-991-8
:GHS08
:GHS07
Storage Temperature:Ambient