
Orthoboric acid ≥99.5%, Reagent Grade
Catalog # B0252-2.5KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Assay (alkalimetric):Min. 99.5 %
- Colour of the solution:complying
- Clearness of the solution:complying
Specification Test Results
About this item
:
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:171 °C
Density:1.510 g/cm³ (20 °C)
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337