Magnesium sulphate, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7487-88-9
:
Formula:MgSO₄
MDL Number:MFCD00011110
:120.37 g/mol
Merck Index:14,5691
Melting Pt:1124 °C (dec.)
Density:2.66
Storage Temperature:Ambient
EINECS:231-298-2
Related Information
Frequently Bought Together