Calcium sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum
CAS Number:7778-18-9
:
Formula:CaO₄S
MDL Number:MFCD00010912
Density:2.964 g/cm³ (20 °C)
Molecular Weight:136.14 g/mol
Melting Pt:1350
Merck Index:13,01711
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-900-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together