

Calcium hydroxide, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E526, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:1305-62-0
:
Formula:Ca(OH)₂
MDL Number:MFCD00010901
:74.09 g/mol
Merck Index:13,01675
Melting Pt:550 °C
Density:2.24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
UN:3262
ADR:8,III
:GHS05
:GHS07
Related Information
Certificates
Search by Lot NumberMERP1.02110.9025_TDS_EN ProductPage_102110
Product specification
msds_102110
SDS
Frequently Bought Together