Calcium hydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7789-78-8
:
Formula:H₂Ca
MDL Number:MFCD00010897
:42.09 g/mol
Merck Index:13,01674
Melting Pt:816 °C
Density:1.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-189-2
UN:1404
ADR:4.3,I
:GHS02
Frequently Bought Together