Order Entry
Austria
ContactUsLinkComponent
 
Calcium hydride ∼ 93%, grains Ø < 2 mm, extra pure
 
undefined
Calcium hydride ∼ 93%, grains Ø < 2 mm, extra pure

 

  • Appearance:
    Grey Powder
  • Assay:
    ≥92 % (Total Ca)
  • Particle size:
    ≤2 mm
  • Chloride (Cl):
    ≤5000 ppm
  • Aluminium (Al):
    ≤100 ppm
  • Hydrogen (H):
    ≥980 ml/g CaH2
  • Iron (Fe):
    ≤100 ppm
  • Magnesium (Mg):
    ≤8000 ppm
  • Nitrogen (N):
    ≤2000 ppm

 

Frequently Bought Together