Strontium carbonate ≥97.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1633-05-2
:
Formula:CO₃Sr
MDL Number:MFCD00011250
Boiling Pt:2647 °C (1 mmHg)
Melting Pt:1100 °C (dec.)
Density:3.70
Molecular Weight:147.63 g/mol
Merck Index:14,8838
Storage Temperature:Ambient
EINECS:216-643-7
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together