Calcium carbonate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, E170, precipitated, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
:100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
Related Information
Certificates
Search by Lot NumberMERP1.02069.1000_TDS_EN MERP1.02069.9025_TDS_EN ProductPage_102069
Product specification
msds_102069
SDS
Frequently Bought Together