Order Entry
Austria
ContactUsLinkComponent
Calcium carbonate, EMSURE® for analysis of silicates, precipitated, Supelco®
Calcium carbonate, EMSURE® for analysis of silicates, precipitated, Supelco®
Catalog # 1.02067.0500
Supplier:  Merck
CAS Number:  
Calcium carbonate, EMSURE® for analysis of silicates, precipitated, Supelco®
Catalog # 1.02067.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (complexometric):
    ≥ 99.0 %
  • Substances insoluble in hydrochloric acid:
    ≤ 0.005 %
  • Chloride (Cl):
    ≤ 0.02 %
  • Sulfate (SO₄):
    ≤ 0.01 %
  • Total nitrogen (N):
    ≤ 0.01 %
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • Ba (Barium):
    ≤ 0.02 %
  • Fe (Iron):
    ≤ 0.001 %
  • K (Potassium):
    ≤ 0.002 %
  • Mg (Magnesium):
    ≤ 0.02 %
  • Na (Sodium):
    ≤ 0.005 %
  • Sr (Strontium):
    ≤ 0.02 %

Specification Test Results

About this item

 :
 :100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C (dec.)
Density:2,71 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906