Order Entry
Austria
ContactUsLinkComponent
Calcium chloride, anhydrous ≥95%, Laboratory Reagent, J.T.Baker®
Irritant
Calcium chloride, anhydrous ≥95%, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Irritant
Calcium chloride, anhydrous ≥95%, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 95 %

Specification Test Results

Related Information

Frequently Bought Together