Sodium sulphate, anhydrous ≥99.0%, granular, ULTRA RESI-ANALYZED® ACS 12-60 Mesh for pesticide residue analysis, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7757-82-6
:
Formula:Na₂SO₄
MDL Number:MFCD00003504
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.68 g/cm³
:142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:≤0.01%
- ACS - Assay (Na₂SO₄):≥99.0 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Loss on Ignition:≤0.5 %
- ACS - pH of 5% Solution at 25°C:5.2 - 9.2
- ACS - Chloride (Cl):≤0.001 %
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Calcium (Ca):≤0.01 %
- ACS - Iron (Fe):≤0.001 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Potassium (K):≤0.01 %
- ACS - Extraction-Concentration Suitability *:Passes Test
- ACS - Heavy Metals (as Pb):≤5 ppm
- Nitrogen Compounds (as N):≤5 ppm
Specification Test Results
Frequently Bought Together