Lithium sulphide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12136-58-2
:
Formula:Li₂S
MDL Number:MFCD00011085
:45.95 g/mol
Boiling Pt:1527 °C (1013 hPa)
Melting Pt:900…975 °C
Density:1.66 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:235-228-1
ADR:4.3,II
:M-Clarity™ quality level = MQ100
UN:2813
Frequently Bought Together