Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate, anhydrous, Suprapur® for trace analysis, Supelco®
Potassium dihydrogen phosphate, anhydrous, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates
Potassium dihydrogen phosphate, anhydrous, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    222 g/l (20 °C)
  • Melting Point:
    253 °C (decomposition)
  • Molar Mass:
    136.08 g/mol
  • Bulk Density:
    1200 kg/m3
  • Density:
    2.34 g/cm3 (20 °C)
  • pH:
    4.4 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together