
Potassium dihydrogen phosphate 98.0-100.5% (dried basis, RT), crystals, GenAR® NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
:GHS07
Specification Test Results
- Bulk Pharmaceutical Chemical:98.0 - 100.5 %
- NF - Assay (KH₂PO₄) (dried basis):98.0 - 100.5 %
- Solubility (5 in 50):Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Arsenic (As):≤3 ppm
- NF - Lead (Pb):≤5 ppm
- NF - Insoluble Substances:≤0.2 %
- NF - Fluoride (F):≤10 ppm
- NF - Loss on Drying at 105°C:≤1.0 %
- NF - Endotoxin Concentration (EU/g):≤5
- NF - pH of a 1 in 100 Solution:4.2 - 4.7
Specification Test Results
Frequently Bought Together