Potassium dihydrogen phosphate, ULTREX®, Ultrapure, J.T.Baker®
4921-04
: Avantor
:
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Pk:100 g
- Seq:3401
:
:136.09 g/mol
:13,07744
:~100 °C (1013 hPa)
:253 °C
:2.34 g/cm³ (20 °C)
:Ambient
:231-913-4
:01-2119490224-41
:GHS07
:7778-77-0
:KH₂PO₄
:MFCD00011401