Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate, BAKER, Laboratory Reagent, J.T.Baker®
Potassium dihydrogen phosphate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # BAKR1737.1000
Supplier:  Avantor
CAS Number:  
Potassium dihydrogen phosphate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # BAKR1737.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    min. 99 %
  • Loss on Drying at 105°C:
    max. 0.1 %

Specification Test Results

About this item

 :
 :136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401