Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate
Potassium dihydrogen phosphate
Catalog # A1043.5000
Supplier:  PanReac AppliChem
CAS Number:  
Potassium dihydrogen phosphate
Catalog # A1043.5000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401