Potassium dihydrogen phosphate 99+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:~ 100 °C (1013 hPa)
Melting Pt:253 °C
Density:2,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals
- Loss on drying:≤1 % (105°C)
- Titration with NaOH:≥99.0 % (on dried substance)
Specification Test Results
Frequently Bought Together