Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate ≥99.0% ACS, Fluka™
Disodium hydrogen phosphate ≥99.0% ACS, Fluka™
Certificates
Disodium hydrogen phosphate ≥99.0% ACS, Fluka™

Specification Test Results

  • pKa (25 °C):
    (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)
  • Assay:
    Min. 99.0 %
  • pH (5 %, 25 °C):
    8.7 - 9.3
  • Loss on drying (105°C):
    Max. 0.2 %
  • Water insoluble matter:
    Max. 0.01 %
  • Iron (Fe):
    Max. 20 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 20 ppm
  • Sulfate (SO₄):
    Max. 50 ppm

Specification Test Results

Related Information

Frequently Bought Together