Order Entry
Austria
ContactUsLinkComponent
di-Potassium hydrogen phosphate ≥96%, TECHNICAL
di-Potassium hydrogen phosphate ≥96%, TECHNICAL
Catalog # 26927.460
Supplier:  VWR Chemicals
CAS Number:  
undefined
di-Potassium hydrogen phosphate ≥96%, TECHNICAL
Catalog # 26927.460
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 96 %

Specification Test Results

About this item

CAS Number:7758-11-4
 :
Formula:K₂HPO₄
MDL Number:MFCD00011383
 :174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5