Order Entry
Austria
ContactUsLinkComponent
di-Potassium hydrogen phosphate, anhydrous 98.0-100.5% (alkalimetric, calculated on anhydrous substance), EMPROVE® api Ph. Eur., BP, USP low sodium, SAFC®
di-Potassium hydrogen phosphate, anhydrous 98.0-100.5% (alkalimetric, calculated on anhydrous substance), EMPROVE® api Ph. Eur., BP, USP low sodium, SAFC®
Catalog # 1.09828.1000
CAS Number:  
undefined
di-Potassium hydrogen phosphate, anhydrous 98.0-100.5% (alkalimetric, calculated on anhydrous substance), EMPROVE® api Ph. Eur., BP, USP low sodium, SAFC®
Catalog # 1.09828.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (alkalimetric, calculated on dried substance):
    98.0 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Insoluble substances:
    ≤ 0.2 %
  • pH-value (5 %; water):
    8.5 - 9.6
  • Carbonate:
    passes test
  • Chloride (Cl):
    ≤ 0.02 %
  • Fluoride (F):
    ≤ 0.001 %
  • Sulfate (SO₄):
    ≤ 0.1 %
  • Heavy metals (as Pb):
    ≤ 0.0010 %
  • Al (Aluminium):
    ≤ 0.0003 %
  • As (Arsenic):
    ≤ 0.0002 %
  • Fe (Iron):
    ≤ 0.0010 %
  • Na (Sodium):
    ≤ 0.1 %
  • Potassium dihydrogen phosphate:
    ≤ 2.5 %
  • Potassium dihydrogen phosphate and tri- Potassium phosphate:
    passes test
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Reducing substances:
    passes test
  • Loss on drying (130 °C):
    ≤ 1.0 %
  • Total aerobic microbial count:
    ≤ 500 CFU/g
  • Bacterial endotoxins:
    < 1.1

Specification Test Results

About this item

 :
 :174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383