di-Kaliumhydrogenphosphat, wasserfrei ≥99,99% (Metall-Basis), Suprapur® für die Spurenanalyse, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7758-11-4
:
Formula:K₂HPO₄
MDL Number:MFCD00011383
:174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2,3 g/cm³ (20 °C)
:M-Clarity™ Qualitätsstufe = MQ100
Storage Temperature:Raumtemperatur
EINECS:231-834-5
Specification Test Results
- Solubility:1600 g/l (20 °C)
- Molar Mass:174.18 g/mol
- Bulk Density:700 - 1000 kg/m3
- Density:2.44 g/cm3 (20 °C)
- pH:9 (10 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Merck Suprapur® High Purity Salts for Elemental Trace Analysis
Product Literature
msds_105109
SDS
ProductPage_105109
Product specification
Frequently Bought Together