Order Entry
Austria
ContactUsLinkComponent
Dipotassium hydrogen orthophosphate trihydrate ≥99%, crystals
Dipotassium hydrogen orthophosphate trihydrate ≥99%, crystals
Supplier:  MP Biomedicals
Certificates
Dipotassium hydrogen orthophosphate trihydrate ≥99%, crystals
Supplier:  MP Biomedicals

Specification Test Results

  • Form:
    Clear Granular Crystals
  • Storage:
    Store at room temperature (15 to 30 °C)

Specification Test Results

Frequently Bought Together