Order Entry
Austria
ContactUsLinkComponent
di-Potassium hydrogen phosphate ≥98.0% (dried basis) FCC, J.T.Baker®
di-Potassium hydrogen phosphate ≥98.0% (dried basis) FCC, J.T.Baker®
Catalog # 3254-05
Supplier:  Avantor
CAS Number:  
undefined
di-Potassium hydrogen phosphate ≥98.0% (dried basis) FCC, J.T.Baker®
Catalog # 3254-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Meets F.C.C. Requirements:
    Passes Test
  • Food GMP Manufactured Product:
    ≥98.0%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.2%
  • Bulk Food Chemical:
    ≤2.0%
  • FCC - Identification:
    Passes Test
  • FCC - Assay (K₂HPO₄) (dried basis):
    ≥98.0 %
  • FCC - Insoluble Substances:
    ≤0.2 %
  • FCC - Loss on Drying:
    ≤2.0 %
  • FCC - Arsenic (As):
    ≤3 ppm
  • FCC - Fluoride (F):
    ≤10 ppm
  • FCC - Lead (Pb):
    ≤2 ppm

Specification Test Results

About this item

 :
 :174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383