Order Entry
Austria
ContactUsLinkComponent
 
di-Potassium hydrogen phosphate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 0242.1000
Supplier:  Avantor
CAS Number:  
undefined
di-Potassium hydrogen phosphate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 0242.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98.0 - 100.5 %
  • Carbonate (CO3):
    passes test
  • Arsenic (As):
    max. 3 ppm
  • Chloride (Cl):
    max. 0.03 %
  • Iron (Fe):
    max. 0.003 %
  • Sodium (Na):
    passes test
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Identification:
    passes test
  • Insoluble Substances:
    max. 0.2 %
  • Limit of fluoride:
    max. 0.001 %
  • Loss on Drying at 105°C:
    max. 1.0 %
  • pH of 5% Solution at 25°C:
    8.5 - 9.6
  • Sulfate (SO4):
    max. 0.1 %

Specification Test Results

About this item

 :
 :174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383