Order Entry
Austria
ContactUsLinkComponent
 
di-Potassium hydrogen phosphate, anhydrous 99+% for analysis
 
undefined
di-Potassium hydrogen phosphate, anhydrous 99+% for analysis
 

 

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤1 % (130°C)
  • pH:
    8.5 to 9.6 (5 % at 20°C)
  • Titration with HClO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤100 ppm
  • Iron (Fe):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

 

 

Frequently Bought Together