Order Entry
Austria
Orders LinkContactUsLinkComponent
Potassium hydroxide

Potassium hydroxide

Corrosive
Irritant
Marketsource
Supplier:   Roth Carl
Potassium hydroxide
Supplier:  Roth Carl
Marketsource
Certificates
Potassium hydroxide

2 Options Available Below

Product Details & Documents
Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Merck Index:12,07806
  • Boiling Pt:1320 °C (1013 hPa)
  • Melting Pt:360 °C
  • Density:2.04 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1813
  • ADR:8,II
  • REACH:01-2119487136-33
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 28,60 €
    28,60 €
    1 * 500 g
       28,60 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 48,70 €
    48,70 €
    1 * 1 kg
       48,70 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.