

Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Merck Index:12,07806
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:2.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
:GHS07
:GHS05
Specification Test Results
- Assay:Min. 85 %
- Total N (Nitrogen):Max. 5 ppm
- Cl (Chloride):Max. 50 ppm
- CO3 (as K2CO3):Max. 2.0 %
- PO4 (Phosphate):Max. 20 ppm
- SiO4 (as SiO2):Max. 100 ppm
- SO4 (Sulphate):Max. 50 ppm
- Al (Aluminium):Max. 10 ppm
- Ca (Calcium):Max. 20 ppm
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
26669_POTASSIUM HYDROXIDE PELLETS GPR RECTAPUR_BSE_REV0
BSE/TSE Certificates
spec_VWRC26669.266 spec_VWRC26669.266 spec_VWRC26669.266 spec_VWRC26669.290 spec_VWRC26669.368
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together