Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide for synthesis
Corrosive
Irritant
Potassium hydroxide for synthesis
Supplier:  Merck
Certificates
Corrosive
Irritant
Potassium hydroxide for synthesis
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1130 g/l (20 °C)
  • Melting Point:
    360 °C
  • Molar Mass:
    56.11 g/mol
  • Boiling Point:
    1320 °C
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.04 g/cm3 (20 °C)
  • pH:
    14 (56 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together