Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide ≥85.0% (by acidimetry), pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, FCC, E525, SAFC®
Potassium hydroxide ≥85.0% (by acidimetry), pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, FCC, E525, SAFC®
Certificates
Potassium hydroxide ≥85.0% (by acidimetry), pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, FCC, E525, SAFC®
Certificates

Specification Test Results

  • Assay (Acidimetric, KOH):
    ≥85.0%
  • Assay (Total Alkalinity Calculated as KOH):
    85.0 - 100.5%
  • Assay (K):
    ≥59.9%
  • Identity:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • Insoluble Substances:
    Passes Test
  • Carbonate (as K₂CO₃):
    ≤2.0%
  • Chloride (Cl):
    ≤0.008%
  • Phosphate (PO₄):
    ≤0.002%
  • Silicate (SiO₂):
    ≤0.01%
  • Sulphate (SO₄):
    ≤0.005%
  • Total nitrogen (N):
    ≤0.0005%
  • Heavy Metals (as Pb):
    ≤0.0005 %
  • Al (Aluminium):
    ≤0.001%
  • As (Arsenic):
    ≤0.0003%
  • Ca (Calcium):
    ≤0.002%
  • Cu (Copper):
    ≤0.0005%
  • Fe (Iron):
    ≤0.001%
  • Hg (Mercury):
    ≤0.00001%
  • Na (Sodium):
    ≤1.0%
  • Pb (Lead):
    ≤0.0002%
  • Zn (Zinc):
    ≤0.0025%
  • Residual Solvents (Ph Eur/USP/ICH):
    Excluded by Manufacturing process

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together