Potassium hydroxide, pellets, EMSURE® ACS, Reag. Ph. Eur. for analysis (max. 0.05% Na), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Merck Index:12,07806
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:2.04 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1813
ADR:8,II
:M-Clarity™ quality level = MQ300
EINECS:215-181-3
REACH:01-2119487136-33
:GHS05
:GHS07
Specification Test Results
- Solubility:1130 g/l (20 °C)
- Melting Point:360 °C
- Molar Mass:56.11 g/mol
- Boiling Point:1320 °C
- Vapor Pressure:(20 °C)
- Density:2.04 g/cm3 (20 °C)
- pH:14 (56 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Pharma Segment Brochure
Product Literature
msds_105029
SDS
ProductPage_105029
Product specification
Frequently Bought Together