Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide ≥85%, powder, Technical Grade, Fluka™
Irritant
Corrosive
Potassium hydroxide ≥85%, powder, Technical Grade, Fluka™
Certificates
Irritant
Corrosive
Potassium hydroxide ≥85%, powder, Technical Grade, Fluka™

Specification Test Results

  • Assay:
    Min. 85 %
  • Iron (Fe):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.1 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 0.002 %
  • Assay of K₂CO₃:
    Max. 1.0 %
  • Vapor Pressure:
    1 mmHg ( 719 °C)

Specification Test Results

Related Information

Frequently Bought Together