

Potassium hydroxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
:1310-58-3
:
:KOH
:MFCD00003553
:56.11 g/mol
:12,07806
:1320 °C (1013 hPa)
:360 °C
:2.04 g/cm³ (20 °C)
:Ambient
:215-181-3
:1813
:8,II
:01-2119487136-33
:GHS05
:GHS07
- Assay:min. 85 %
- Aluminium (Al):max. 0.001 %
- Calcium (Ca):max. 0.001 %
- Sodium (Na):max. 0.5 %
- Potassium Carbonate (K2CO3):max. 1.0 %
- Silica (SiO2):max. 0.005 %
- Trace Impurities (in ppm) Chloride (Cl):max. 5
- Trace Impurities (in ppm) Iron (Fe):max. 5
- Trace Impurities (in ppm) Nickel (Ni):max. 5
- Trace Impurities (in ppm) Heavy Metals (as Pb):max. 5
- Trace Impurities (in ppm) Nitrogen Compounds (as N):max. 3
- Trace Impurities (in ppm) Phosphate (PO4):max. 5
- Trace Impurities (in ppm) Sulfate (SO4):max. 5
Frequently Bought Together